Implant Line
MED-4535
HIGH CONSISTENCY SILICONE ELASTOMER
Description
  • Uncatalyzed, vinyl-functional, high consistency silicone elastomer
  • Low durometer silicone
  • Requires the addition of a peroxide catalyst to accomplish vulcanization
  • Strained through a 400-mesh screen (minimum)
Applications
  • For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
NuSil Technology’s MED-4535 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer 35 Type A
Tensile 1,300 psi (8.97 MPa)
Elongation 800 %
Tear 125 ppi (22.05 kN/m)
Appearance Translucent
Cure 5 minutes / 116 °C
Cure System Peroxide
Mix Ratio 1 PART
Plasticity
Post Cure 2 hours / 177 °C
Specific Gravity 1.1
Stress at Strain 185 psi / 200 % (1.28 MPa / 200 %)
Comment Uncatalyzed
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.