Implant Line
MED-4211
LOW VISCOSITY SILICONE ELASTOMER
Description
  • A two-part, translucent pourable silicone elastomer
  • Cures with heat via addition-cure chemistry
  • 10:1 Mix Ratio (Part A: Part B)
Applications
  • For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property Average Result
Durometer 25 Type A
Viscosity 97,800 cP (97,800 MPa*s)
Tensile 685 psi (4.72 MPa)
Appearance Translucent
Cure 3 minutes / 150 °C
Cure System Platinum
Elongation 530 %
Mix Ratio 10:1
Post Cure 1 hour / 150 °C
Specific Gravity 1.1
Comment Flowable
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.