Premium Care Line
MED-4032
HIGH CONSISTENCY SILICONE ELASTOMER
Description
  • A unique three-part system, VersaSil3 offers the flexibility of adjustable cure rate and table life for various fabrication requirements
  • High tear strength, wide processing parameters, and translucent, non-tacky surfaces
  • Developed to be compounded with the inhibitor CAT-40 and the platinum catalyst CAT-55. Advantages include: no volatile by-products, lower cure temperature, and optional post-cure
  • 100: 1.0: 1.0 Mix Ratio
Applications
  • For mass production with maximum flexibility
  • For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
NuSil Technology’s VersaSil3 Series shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer 30 Type A
Tensile 1,200 psi (8.28 MPa)
Elongation 1,100 %
Tear 160 ppi (28.22 kN/m)
Cure 10 minutes / 116 °C
Cure Rate Scorch 1 minutes
Cure Rate T90 3.05 minutes
Mix Ratio 100:1:1
Special Feature
Specific Gravity 1.1
Stress at Strain 140 psi / 200 % (0.97 MPa / 200 %)
Work Time 8 hours
Comment Can be Cured via Platinum or Peroxide
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.