Premium Care Line
MED-2045
HIGH CONSISTENCY SILICONE ELASTOMER
Description
  • A series of color masterbatches consisting of pigments dispersed in a vinyldimethyl-terminated polydimethylsiloxane polymer
  • Consistency of a low viscosity paste
  • Supported by USP Class VI and ISO 10993 Biological testing (reference Biological Testing Data Table)
Applications
  • For easy and more precise pigmentation of LSR and low consistency materials, which may be injection molded
  • Eliminates use of powders that may contaminate clean room environments
  • Reduces production time
  • Strict quality controls ensure color consistency
  • Homogeneity of masterbatch minimizes agglomerates
NuSil Technology’s MED-2045 shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer 40 Type A
Tensile 1,500 psi (10.34 MPa)
Elongation 800 %
Tear 200 ppi (35.27 kN/m)
Appearance Translucent
Cure 10 minutes / 171 °C
Cure Rate Scorch 0.7 minutes
Cure Rate T90 2.35 minutes
Cure System Platinum
Mix Ratio 100:0.7:0.16
Plasticity
Post Cure 2 hours / 148 °C
Specific Gravity 1.16
Stress at Strain 200 psi / 200 % (1.38 MPa / 200 %)
Comment Designed for Dissolving in Solvents
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.