Description
- Specialty formulated primer
- One-component primer supplied in VM&P Naphtha requires no mixing
- Easy to use, air-drying, one-part primer designed for use with platinum-cured systems on polycarbonates
- Flash Point (TCC) of 17°C (63°F)
- Convenient container sizes yield less waste
Applications
- Improves adhesion of silicones to various substrates including: metals (such as stainless steel, steel, copper and aluminum), ceramics, rigid plastics, and other silicone materials
NuSil Technology’s MED-162 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Cure | 2 hours / RTV |
Mix Ratio | 1 PART |
Non Volatile Content | 15 % |
Specific Gravity | 0.79 |
Appearance | Translucent |
Solvent | Naptha |
Comment | Increased adhesion to polycarbonate (PC), polyurethane (PU) and polyetheretherketone (PEEK). |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.