Description
- Specially formulated primer
- One-component primer supplied in VM&P Naphtha requires no mixing
- Easy to use, air-drying, one-part primer designed for use with one and two-part RTV systems where conventional silicone primers are insufficient
- Flash Point (TCC) of 17°C (63°F)
- Convenient container sizes yield less waste
Applications
- Improves adhesion of silicones to various substrates including: metals (such as stainless steel, steel, copper and aluminum), ceramics, rigid plastics, and other silicone materials
Product Details
| Property | Average Result |
|---|---|
| Cure | 1 hours / RTV |
| Mix Ratio | 1 PART |
| Non Volatile Content | 4 % |
| Specific Gravity | 0.77 |
| Appearance | Translucent |
| Lap Shear | 330 psi (2.28 MPa) |
| Solvent | Naptha |
| Comment | General all-purpose primer. Use with platinum or moisture cure silicones. |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.