Description
- One-part, non-curing, pressure sensitive adhesive
Applications
- For temporarily adhering various substrates (including metals, glass, silicone, organic rubbers, and skin)
- Use for medical tapes and fasteners, external incontinence devices, transdermal skin patches, hair replacement adhesives, and special effects adhesives
- Non-irritating to skin
NuSil Technology’s MED-1356 shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Peel Strength | 14.3 lbf/in (2.5 kN/m) |
Viscosity | 250 cP (250 mPa*s) |
Non Volatile Content | 50 % |
Solvent | Ethyl Acetate |
Cure System | Platinum |
Mix Ratio | 1 PART |
Tack | 2.3 psi (0.02 MPa) |
Comment | LOW MODULUS, HIGH TACK |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.