Description
- One part, low slump, solvent-free translucent silicone
- Cures at room temperature upon exposure to atmospheric moisture
Applications
- Well suited for use as an adhesive for bonding and sealing silicone materials
- For applications requiring the bonding of silicone to metals, urethanes and various other substrates
NuSil Technology’s MED-1137 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
| Property | Average Result |
|---|---|
| Cure Type | RTV |
| Mix Ratio | 1 PART |
| Rheology | Thixotropic / non-slump |
| Appearance | Translucent |
| Cure | 72 hours / RTV |
| Cure System | Acetoxy |
| Durometer | 30 Type A |
| Elongation | 560 % |
| Extrusion Rate | 180 g/minute |
| Flow Rate | 0.2 in (5.08 mm) |
| Specific Gravity | 1.07 |
| Stress at Strain | 170 psi / 200 % (1.17 MPa / 200 %) |
| Substrate | Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium / With Primer: Polycarbonate |
| Tack Free Time | 9 minutes |
| Tensile | 700 psi (4.83 MPa) |
| Comment | Low-slump, solvent-free translucent silicone |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.