Description
- One part, low slump, solvent-free translucent silicone
- Cures at room temperature upon exposure to atmospheric moisture
Applications
- Well suited for use as an adhesive for bonding and sealing silicone materials
- For applications requiring the bonding of silicone to metals, urethanes and various other substrates
Product Details
| Property | Average Result |
|---|---|
| Cure Type | RTV |
| Mix Ratio | 1 PART |
| Rheology | Thixotropic / non-slump |
| Appearance | Translucent |
| Cure | 72 hours / RTV |
| Cure System | Acetoxy |
| Durometer | 30 Type A |
| Elongation | 560 % |
| Extrusion Rate | 180 g/minute |
| Flow Rate | 0.2 in (5.08 mm) |
| Specific Gravity | 1.07 |
| Stress at Strain | 170 psi / 200 % (1.17 MPa / 200 %) |
| Substrate | Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium / With Primer: Polycarbonate |
| Tack Free Time | 9 minutes |
| Tensile | 700 psi (4.83 MPa) |
| Comment | Low-slump, solvent-free translucent silicone |
Contact NuSil® to Order Now
For nearly 40 years, the NuSil brand has offered a wide range of high-purity silicone adhesives for use in medical devices. Our team is ready to collaborate with you to provide medical grade silicone solutions to satisfy your unique adhesion requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.