Description
- One part, low slump, solvent-free translucent silicone
- Cures at room temperature upon exposure to atmospheric moisture
Applications
- Well suited for use as an adhesive for bonding and sealing silicone materials
- For applications requiring the bonding of silicone to metals, urethanes and various other substrates
NuSil Technology’s MED-1131 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
| Property | Average Result |
|---|---|
| Cure Type | RTV |
| Mix Ratio | 1 PART |
| Rheology | Thixotropic / non-slump |
| Appearance | Translucent |
| Cure | 7 days / RTV |
| Cure System | Oxime |
| Durometer | 35 Type A |
| Elongation | 330 % |
| Flow Rate | 0.4 in (10.16 mm) |
| Specific Gravity | 1.1 |
| Substrate | Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium |
| Tack Free Time | 20 minutes |
| Tear | 45 ppi (7.94 kN/m) |
| Tensile | 890 psi (6.14 MPa) |
| Comment | Non-acidic leaving group |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.