Description
- High purity, optically clear, firm silicone gel
- Two medium viscosity components blend easily in a convenient 1:1 ratio by weight or volume
Applications
- For potting, encapsulating, backfilling, and dampening applications requiring a firm gel with optical clarity
- Recommended for use with automatic dispense systems
- For applications requiring an operating temperature range of
- -65°C to 200°C (-85°F to 392°F)
Product Details
Property | Average Result |
---|---|
Viscosity | 14,500 cP (14,500 mPa*s) |
Penetration | 0.4 mm |
Appearance | Clear |
Cure | 30 minutes / 150 °C |
Cure System | Platinum |
Mix Ratio | 1:1 |
Operating Temperature | -65 °C / 200 °C |
Refractive Index | 1.41 |
Comment | Soft Low Tack Gel; Tough gel and 24 hr minimum potlife based on 2X initial viscosity |
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.