GEL-8246
FIRM SILICONE GEL
Description
  • 1 : 1 Mix Ratio (Part A : Part B)
  • High purity, higher viscosity, clear silicone gel
  • RoHS/REACh compliant
Applications
  • For protection of sensitive assemblies from mechanical shock, thermal shock, dust, and atmospheric contamination
  • For applications requiring optically clear materials with a refractive index of 1.47 at 589nm
  • Higher viscosity allows reduced flow into unwanted areas
Product Details
Property Average Result
Viscosity 35,000 cP (35,000 MPa*s)
Appearance Clear
Durometer 55 Type 00
Refractive Index 1.47
Specific Gravity 1.05
Volatile Content 0.4 %
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.