Description
- Clear, tacky silicone gel
- Easy mixing two-component, low viscosity system
- 1:1 Mix Ratio (Part A:B)
- Platinum Cure
Applications
- Ideal for casting, calendering, and backfilling.
- Suitable for applications requiring a clear, tacky, firm silicone gel
- For use as an embedding or potting compound for protection of electronic assemblies and components from environmental contamination
- Provide excellent stress relief for thermal cycling of sensitive components such as wire bonds and thin wafers
- Electrically insulating and non-conductive
- Ideal for potting and filling intricate assemblies, LCD bonding, and has excellent adhesion at various thickness for tapes.
- Ideal for use in automated dispensing equipment
Product Details
Property | Average Result |
---|---|
Viscosity | 450 cP (450 mPa*s) |
Penetration | 14 mm |
Appearance | Clear |
Cure | 60 minutes / 140 °C |
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.