GEL-8132
TACKY SILICONE GEL
Description
  • Clear, tacky, firm silicone gel
  • Easy mixing two-component, medium viscosity system
  • 1:1 Mix Ratio (Part A:B)
Applications
  • Ideal for casting, calendering, backfilling and dampening
  • Suitable for applications requiring a clear, tacky, firm silicone gel
  • Consider for the use as the tacky substrate for removable tapes.
Product Details
Property Average Result
Viscosity 12,000 cP (12,000 MPa*s)
Penetration 1.1 mm
Appearance Translucent
Cure System Platinum
Mix Ratio 1:1
Comment High tack, low penetration, for use as adhesive layer on removable tacky films and tapes. (5 seconds)
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.