GEL-8130
TACKY SILICONE GEL
Description
  • Clear, tacky, firm silicone gel
  • A medium viscosity liquid that cures to a soft silicone
  • 1:1 Mix Ratio (Part A:B)
Applications
  • Ideal for casting, calendering, backfilling and dampening
  • Suitable for applications requiring a clear, tacky, firm silicone gel
  • Consider for the use as the tacky substrate for removable tapes.
Product Details
Property Average Result
Viscosity 25,200 cP (25,200 mPa*s)
Penetration 2.2 mm
Work Time 2.5 hours
Appearance Translucent
Cure 30 minutes / 100 °C
Cure System Platinum
Mix Ratio 1:1
Volatile Content 1.4 %
Comment Medium tack, low penetration, for use as adhesive layer on removable tacky films and tapes. (5 seconds)
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.