Description
- Pre-catalyzed, high consistency elastomer (HCR)
- Strained through a 400-mesh screen (minimum) to ensure freedom from particulate contamination
Applications
- Applications requiring solvent resistance
- Transfer and compression molding parts such as: o-rings, gaskets, stoppers and seals
- Calendering and extruding
Product Details
Property | Average Result |
---|---|
Cure System | Peroxide |
Durometer | 30 Type A |
Tensile | 850 psi (5.86 MPa) |
Elongation | 300 % |
Tear | 40 ppi (7.05 kN/m) |
Appearance | Translucent |
Cure | 30 minutes / 120 °C |
Plasticity | |
Post Cure | 1 minutes / 150 °C |
Specific Gravity | 1.33 |
Comment | HCR Fluoro Elastomer;Recommend for long term fuel and solvent exposure. Can be compression molded and requires heat to cure. High tear. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.