FS-3606
SILICONE FLUID
Description
  • A clear liquid copolymer consisting of dimethylsiloxane and trifluoropropylmethylsiloxane
  • Available in standard viscosities of 350, 1,000 and 12,500 cP and custom viscosities upon request
  • Has reduced solubility, good wetting properties and good lubrication characteristics for plastics, rubber and skin
  • Highly water repellent and resists decomposition by heat and oxidation
Applications
  • To provide a lubricious and/or hydrophobic coating
  • For lubricating the surfaces of molded dimethyl silicone elastomer parts
  • For applications requiring an operating temperature range of -40°C to 200°C (-40°F to 392°F)
Product Details
Property Average Result
Viscosity 12,500 cP / 1,000 cP / 350 cP (12,500 mPa*s / 1,000 mPa*s / 350 mPa*s)
Volatile Content 2 %
Specific Gravity 1.28
Appearance Translucent
Dielectric Strength 400 volts/mil (15.6 kV/mm)
Operating Temperature -40 °C / 200 °C
Refractive Index 1.38
Volume Resistivity 1,000,000,000,000,000 ohmcm
Comment 60%m Fluoro polymer.
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.