Description
- Two-part, translucent, fluorosilicone elastomer used with liquid injection molding equipment
- 1:1 Mix Ratio (Part A: Part B)
- Cures rapidly with heat
- 100 mol % fluorosilicone
Applications
- For components exposed to JP-8, aviation fuels and solvents
- For applications requiring a medium hardness silicone with solvent resistance
- Some typical applications include O-rings, gaskets, seals, and precision molded parts
- Excellent for molding high quality parts at rapid rates
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Durometer | 40 Type A |
Tensile | 1,150 psi (7.93 MPa) |
Elongation | 335 % |
Tear | 60 ppi (10.58 kN/m) |
Appearance | Translucent |
Cure | 30 minutes / 150 °C |
Extrusion Rate | 35 g/minute |
Mix Ratio | 1:1 |
Specific Gravity | 1.39 |
Work Time | 24 hours |
Comment | LSR Fluoro Elastomer;For long term fuel or solvent exposure. Can be liquid injection molded and requires heat to cure. Recommend SP-120 or SP-201 |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.