Description
- A two-part, white, fluorosilicone gel
- 100 mol% fluorosilicone
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For filling of small gaps
- For applications requiring solvent and/or fuel resistance
- Bonds aggressively to most surfaces
- For components exposed to JP-8, aviation fuels and solvents
Product Details
Property | Average Result |
---|---|
Viscosity | 1,200 cP (1,200 mPa*s) |
Appearance | White |
Cure | 4 hours / 50 °C |
Cure System | Platinum |
Durometer | 10 Type 00 |
Mix Ratio | 1:1 |
Comment | Firm Fluoro Gel;Recommended for long term solvent / fuel exposure, 6,000 cP maximum mixed viscosity after 3 hrs |
Making the right choice for fuel- and solvent-resistant silicones
Learn how high-purity fluorosilicones resist degradation and maintain performance in aviation and defense applications.