Applications
- Adhesives for bonding and sealing silicones to each other and other substrates such as metals and plastics such as polycarbonate, and PET.
- Protects electrical components and assemblies against shock, vibration, moisture, dust, chemicals, and other environmental hazards
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Extrusion Rate | 0.05 g/minute |
Durometer | 28 Type A |
Appearance | Translucent |
Cure | 10 minutes / 116 °C |
Elongation | 540 % |
Specific Gravity | 1.09 |
Tear | 90 ppi (15.87 kN/m) |
Tensile | 830 psi (5.72 MPa) |
Work Time | 2 hours |
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.