EPM-2890
THERMALLY CONDUCTIVE NON-CORROSIVE RTV SILICONE ADHESIVE
Description
  • One-Part, white RTV silicone
  • Non-slumping in thin sections
Applications
  • To provide moderate heat transfer between electrical/electronic components and their heat sinks
  • As a sealing, caulking, adhesive or potting material in electronics applications requiring minimal outgassing to avoid condensation in sensitive devices
  • Use for any application where grooves and other configurations require a non-slumping, non-flowable to limited flow one-part material
  • For applications requiring a broader operating temperature range
Product Details
Property Average Result
Thermal Conductivity 0.61 W/(mK)
Extrusion Rate 40 g/minute
Cure System Oxime
Tensile 400 psi (2.76 MPa)
Appearance White
Cte
Cure 72 hours / RTV
Durometer 65 Type A
Elongation 150 %
Ionic Content Cl <5 ppm
Ionic Content K <5 ppm
Ionic Content Na <5 ppm
Lap Shear 225 psi (1.55 MPa)
Mix Ratio 1 PART
Specific Gravity 2.33
Tack Free Time 40 minutes
Tear 55 ppi (9.7 kN/m)
Comment Excellent for bonding in thin layers and does not contain Boron Nitride.
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.