Description
- One-Part, RTV Condensation CureOxime CureNon-SlumpRoHS compliant
Applications
- For applications requiring low volatility and high purity
- Designed for protection of electrical components and assemblies against shock, vibration, moisture, dust, chemicals, and other environmental hazards
- For bonding and sealing in applications such as overhead or vertical joints that require a non-slumping and one-part material
- Bonds well to plastics such as polycarbonate
Product Details
Property | Average Result |
---|---|
Durometer | 35 Type A |
Appearance | Translucent |
Dielectric Strength | 1,100 volts/mil (42.9 kV/mm) |
Elongation | 280 % |
Flow Rate | <2 in (<50.8 mm) |
Specific Gravity | 1.11 |
Tack Free Time | 25 minutes |
Tensile | 685 psi (4.72 MPa) |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.