Description
- Two-part, white, thermally conductive silicone
- Cures with the addition of heat
- Designed to allow bond line thicknesses - 50 micron to reduce thermal resistance
- 20:1 Mix Ratio (Part A: Part B)
Applications
- For applications requiring low volatility under extreme operating conditions to avoid contamination in sensitive devices
- To provide heat transfer between electrical/electronic components and their heat sinks while remaining electrically insulating
- Use to adhere integrated circuit substrates, base plates, heat sinks or where grooves or other configurations require a non-flowable to limited flow material
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 0.64 W/(mK) |
Extrusion Rate | 140 g/minute |
Cure System | Platinum |
Tensile | 400 psi (2.76 MPa) |
Appearance | White |
Cure | 30 minutes / 150 °C |
Durometer | 55 Type A |
Elongation | 225 % |
Ionic Content Cl | <6 ppm |
Ionic Content K | <3 ppm |
Ionic Content Na | <3 ppm |
Mix Ratio | 20:1 |
Tear | 55 ppi (9.7 kN/m) |
Work Time | 3 hours |
Comment | Recommended for bondlines 50 micron or greater. (Contains Boron Nitride) |
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.