Description
- Two-part, clear silicone gel
- 1:1 Mix Ratio
- (Part A: Part B)
Applications
- For applications requiring low volatility
- Useful for potting intricate assemblies due to a low viscosity
- As an embedding or potting compound for environmental protection of electronic assemblies and components
Product Details
Property | Average Result |
---|---|
Viscosity | 2,500 cP (2,500 mPa*s) |
Penetration | 4 mm |
Work Time | 30 hours |
Appearance | Clear |
Cure | 30 minutes / 150 °C |
Cure System | Platinum |
Ionic Content Cl | <5 ppm |
Ionic Content K | <1 ppm |
Ionic Content Na | <2 ppm |
Mix Ratio | 1:1 |
Volume Resistivity | 100,000,000,000,000 ohmcm |
Comment | Soft High Tack Gel |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.