Description
- Two-part, tan-colored, electrically conductive silicone
- 20:1 Mix Ratio (Part A: Part B)
Applications
- For applications requiring low volatility
- Well suited for form-in-place conductive gaskets
- Use as a thermally or electrically conductive interface material
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 186,200 cP (186,200 mPa*s) |
Durometer | 85 Type A |
Appearance | Tan |
Cte | |
Cure | 30 minutes / 150 C |
Elongation | 70 % |
Ionic Content Cl | <5 ppm |
Ionic Content K | <7 ppm |
Ionic Content Na | <5 ppm |
Mix Ratio | 20:1 |
Specific Gravity | 3.39 |
Tensile | 525 psi (3.62 MPa) |
Thermal Conductivity | 1.2 W/(mK) |
Volume Resistivity | 0.01 ohmcm |
Work Time | 2.75 hours |
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.