Description
- Two-part, black, static dissipative silicone
- Offers low modulus
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For applications requiring low volatility
- Use for static dissipation in electronic components
- Use for any application where grooves and other configurations require a non-flowing to limited flow material
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 675,000 cP (675,000 mPa*s) |
Lap Shear | 325 psi (2.24 MPa) |
Durometer | 30 Type A |
Appearance | Black |
Cure | 24 hours / RTV |
Elongation | 350 % |
Ionic Content Cl | <5 ppm |
Ionic Content K | <1 ppm |
Ionic Content Na | <6 ppm |
Mix Ratio | 1:1 |
Operating Temperature | -100 °C / 300 °C |
Specific Gravity | 1.05 |
Tear | 30 ppi (5.29 kN/m) |
Tensile | 500 psi (3.45 MPa) |
Thermal Conductivity | 0.22 W/(mK) |
Volume Resistivity | 535 ohmcm |
Work Time | 1 minutes |
Comment | Carbon filled. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.