Description
- 1:1 Mix ratio (Part A: Part B), Platinum addition cure
- Controlled Aspect Ratio
- Translucent
- Cures at room temperature
- Low Volatility
- RoHS compliant
Applications
- Applications that require non-slumping material
- Cures quickly to form cure in place gasket in device
- Can be easily removed after cure to reduce rework time
- Leaves minimal to no residue once removed
- Easily dispensed and consistent aspect ratio
- Suitable for mix and meter equipment
Product Details
| Property | Average Result |
|---|---|
| Cure System | Platinum |
| Extrusion Rate | 0.08 g/minute |
| Durometer | 30 Type A |
| Appearance | Translucent |
| Cte | |
| Cure | 24 hours / RTV |
| Dielectric Strength | 530 volts/mil (20.67 kV/mm) |
| Elongation | 450 % |
| Tensile | 850 psi (5.86 MPa) |
| Thermal Conductivity | 0.18 W/(mK) |
| Volume Resistivity | 100,000,000,000,000 ohmcm |
| Work Time | 20 minutes |
| Comment | Mobile Devices, Displace |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.