Description
- One-part, black, platinum catalyzed silicone glob-top
- Thixotropic consistency
- Low modulus
Applications
- For applications requiring low volatility
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ensures microchip reliability by providing vital protection
- For applications that require precise delivery configurations
Product Details
| Property | Average Result |
|---|---|
| Cure System | Platinum |
| Viscosity | 300,000 cP (300,000 mPa*s) |
| Extrusion Rate | 0.9 g/minute |
| Durometer | 20 Type A |
| Appearance | Black |
| Cure | 15 minutes / 200 °C |
| Elongation | 700 % |
| Mix Ratio | 1 PART |
| Specific Gravity | 1.15 |
| Stress at Strain | 50 psi / 100 % (0.34 MPa / 100 %) |
| Tear | 50 ppi (8.82 kN/m) |
| Tensile | 750 psi (5.17 MPa) |
| Volume Resistivity | 1,000,000,000,000,000 ohmcm |
| Comment | Low volatility, Thixotropic, dispensable and 0.5 mm maximum slump. Shipped and stored frozen. |
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.