EPM-2410-2
LOW VOLATILITY SILICONE ADHESIVE
Description
  • Two-part, black silicone system Offers a high tear strength 1:1 Mix Ratio (Part A: Part B)
Applications
  • To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength For use as an adhesive
Product Details
Property Average Result
Viscosity 50,000 cP (50,000 mPa*s)
Lap Shear 425 psi (2.93 MPa)
Durometer 30 Type A
Appearance Black
Cure 15 minutes / 150 °C
Elongation 400 %
Ionic Content Cl <5 ppm
Ionic Content K <1 ppm
Ionic Content Na <1 ppm
Tack Free Time 5 hours
Tensile 750 psi (5.17 MPa)
Volatile Content 0.60 %
Work Time 30 minutes
Bonding
The diverse line of NuSil® adhesives specifically designed for electronic component assembly increases production throughput without sacrificing performance.