Description
- Two-part, black silicone system Offers a high tear strength 1:1 Mix Ratio (Part A: Part B)
Applications
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength For use as an adhesive
Product Details
| Property | Average Result |
|---|---|
| Viscosity | 50,000 cP (50,000 mPa*s) |
| Lap Shear | 425 psi (2.93 MPa) |
| Durometer | 30 Type A |
| Appearance | Black |
| Cure | 15 minutes / 150 °C |
| Elongation | 400 % |
| Ionic Content Cl | <5 ppm |
| Ionic Content K | <1 ppm |
| Ionic Content Na | <1 ppm |
| Tack Free Time | 5 hours |
| Tensile | 750 psi (5.17 MPa) |
| Volatile Content | 0.60 % |
| Work Time | 30 minutes |
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.