Product Details
| Property | Average Result |
|---|---|
| Viscosity | 50,000 cP (50,000 mPa*s) |
| Lap Shear | 400 psi (2.76 MPa) |
| Durometer | 30 Type A |
| Appearance | White |
| Cure | 15 minutes / 150 °C |
| Elongation | 350 % |
| Ionic Content Cl | <15 ppm |
| Ionic Content K | <1 ppm |
| Ionic Content Na | <2 ppm |
| Tack Free Time | 4 hours |
| Tensile | 700 psi (4.83 MPa) |
| Thermal Conductivity | 0.23 W/(mK) |
| Volatile Content | 0.5 % |
| Work Time | 30 minutes |
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.