Product Details
| Property | Average Result |
|---|---|
| Viscosity | 50,000 cP (50,000 mPa*s) |
| Lap Shear | 400 psi (2.76 MPa) |
| Durometer | 30 Type A |
| Appearance | White |
| Cure | 15 minutes / 150 °C |
| Elongation | 350 % |
| Ionic Content Cl | <15 ppm |
| Ionic Content K | <1 ppm |
| Ionic Content Na | <2 ppm |
| Tack Free Time | 4 hours |
| Tensile | 700 psi (4.83 MPa) |
| Thermal Conductivity | 0.23 W/(mK) |
| Volatile Content | 0.5 % |
| Work Time | 30 minutes |
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.