EPM-2401
LOW VOLATILITY THERMAL INTERFACE SILICONE GREASE
Description
  • Thermally conductive, white silicone grease
  • Excellent resistance to moisture, ozone and oxidation over a wide temperature range
Applications
  • For applications requiring low volatility
  • For moderate transfer of heat, high and low temperature stability, and low bleed
Product Details
Property Average Result
Thermal Conductivity 0.7 W/(mK)
Cure System Non-Curing
Appearance White
Ionic Content Cl <5 ppm
Ionic Content K <2 ppm
Ionic Content Na <4 ppm
Specific Gravity 2.3
Comment Versatile, low volatility, thermally conductive grease where thin bondlines or a compressible grease is needed. Non-curing.
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.