Description
- High clarity silicone
- High purity; <0.05% D4 cyclic content
- Lower process viscosity for ease of use
- Proprietary process creates an elastomer gel free of pilling and balling
- Delivers a silky and non–greasy feel within skin care applications
- Increased shine and gloss
- Ideal for non-irritating and sensitive skin care applications
- D6 offers a unique sensory feel in formulations
Applications
- Moisturization
- Anti-Aging/Anti-Wrinkle
- Sunscreen, After- Sun, and Self- Tanners
- Skin Repair Serums
- Skin Tone Repair
- Men’s Grooming
- Color Cosmetics
Product Details
Property | Average Result |
---|---|
INCI Name | Cyclopentasiloxane (and) Cyclohexasiloxane (and) Dimethicone/Vinyl Dimethicone Crosspolymer |
D4 Content | <0.05 % |
Viscosity | 60,000 cP (60,000 mPa*s) |
Appearance | Clear |
Non Volatile Content | 8 % |
Refractive Index | 1.40 |
Specific Gravity | 0.96 |
Comment | 8% Solids Elastomer Gel Dispersed in D5 and D6 |