CV-7300
CONTROLLED VOLATILITY SILICONE FLUID
Description
  • A clear, dimethyl polysiloxane
  • Available in viscosities of 100; 350; 500; 1,000; 2,000; 5,000; 10,000; 12,500; 30,000; 50,000; 100,000; and 300,000 cP
Applications
  • For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
  • For vibration and dampening control
  • As working media in fluid drive systems
Meets or exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤1% and CVCM of ≤0.1%
Product Details
Property Average Result
Viscosity 100 cP / 350 cP / 500 cP / 1,000 cP / 2,000 cP / 5,000 cP / 10,000 cP / 12,500 cP / 30,000 cP / 50,000 cP / 100,000 cP / 300,000 cP
Specific Gravity 0.97
Appearance Clear
Refractive Index 1.40
Comment Refractive Index 1.40
High-precision laser performance with ultra-low outgassing silicone
Learn how NuSil collaborated with a leading manufacturer of lasers to improve longevity and stability of space-based lasers.