Description
- Two-part, white, thermally conductive RTV silicone
- 100:0.2 Mix Ratio (Base: Curing Agent)
- Designed for enhanced performance in extreme low and high temperatures
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- To provide heat transfer between electrical/electronic components and their heat sinks
- Use to adhere integrated circuit substrates, base plates, heat sinks or where grooves or other configurations require a non-flowable to limited flow material
- For applications requiring a broader operating temperature range
Product Details
| Property | Average Result |
|---|---|
| Thermal Conductivity | 1.95 W/(mK) |
| Cure System | Alkoxy |
| Tensile | 250 psi (1.72 MPa) |
| Appearance | White |
| Cure | 7 days / RTV |
| Durometer | 80 Type A |
| Elongation | 30 % |
| Lap Shear | 150 psi (1.03 MPa) |
| Mix Ratio | 100:0.2 |
| Specific Gravity | 1.57 |
| Tear | 45 ppi (7.94 kN/m) |
| Work Time | 2.5 hours |
| Comment | *22) 2.0 W/mK, *3) Primed Lap Shear 150 psi (1.0 MPa) |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.