Description
- Two-part, gray, thermally conductive silicone
- Based on a dimethyl silicone polymer
- Uses a platinum-catalyzed addition cure
- 20:1 Mix Ratio (Part A:B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- To provide moderate heat transfer between electrical/electronic components and their heat sinks
- Use to adhere openings in modules or housings where grooves and other configurations require a non-flowable to limited flow material
Meets or exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤1% and CVCM of ≤0.1%
Product Details
| Property | Average Result |
|---|---|
| Thermal Conductivity | 1 W/(mK) |
| Cure System | Platinum |
| Tensile | 650 psi (4.48 MPa) |
| Appearance | Gray |
| Cte | |
| Cure | 24 hours / RTV |
| Dielectric Strength | 430 volts/mil (16.77 kV/mm) |
| Durometer | 85 Type A |
| Elongation | 15 % |
| Lap Shear | 375 psi (2.59 MPa) |
| Mix Ratio | 20:1 |
| Specific Gravity | 2.4 |
| Tack Free Time | 4 hours |
| Tear | 55 ppi (9.7 kN/m) |
| Volume Resistivity | 140,000,000,000,000 ohmcm |
| Work Time | 2.5 hours |
| Comment | *22) 1.0 W/mK, *4) Primed Lap Shear 375 psi (2.6 MPa) |
What Is Space Grade?
Discover what it takes to make silicones worthy of the distinction "Space Grade" and the rigorous testing that these materials undergo.