Description
- One-Part, white RTV silicone
- Non-slumping in thin sections
- Non-corrosive
- Designed for enhanced performance in extreme low and high temperatures
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- To provide moderate heat transfer between electrical/electronic components and their heat sinks
- As a sealing, caulking, adhesive or potting material in electronics and space applications requiring minimal outgassing to avoid condensation in sensitive devices
- For bonding and sealing in applications such as overhead or vertical joints that require a non-slumping and one-part material
- For applications requiring a broader operating temperature range
Product Details
| Property | Average Result |
|---|---|
| Thermal Conductivity | 0.61 W/(mK) |
| Extrusion Rate | 40 g/minute |
| Cure System | Oxime |
| Tensile | 400 psi (2.76 MPa) |
| Appearance | White |
| Cte | |
| Cure | 72 hours / RTV |
| Durometer | 65 Type A |
| Elongation | 150 % |
| Ionic Content Cl | <5 ppm |
| Ionic Content K | <5 ppm |
| Ionic Content Na | <5 ppm |
| Mix Ratio | 1 PART |
| Specific Gravity | 2.33 |
| Tack Free Time | 40 minutes |
| Comment | *22) 0.6 W/mK, Low Temperature |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.