Description
- A clear, two-part, pourable RTV silicone
- 100: 0.5 Mix Ratio (Base: Curing Agent)
Applications
- For applications requiring low outgassing and minimal volatile condensibles under extreme operating conditions to avoid condensation in sensitive devices
- As a sealing, caulking, adhesive or potting material in electronic and space applications where high vacuum environments are encountered
- As a diluent for reducing the viscosity of other tin condensation controlled volatility silicones because of its low viscosity
- For applications requiring a broader operating temperature range
Product Details
| Property | Average Result |
|---|---|
| Cure System | Alkoxy |
| Viscosity | 5,000 cP (5,000 mPa*s) |
| Lap Shear | 45 psi (0.31 MPa) |
| Durometer | 21 Type A |
| Appearance | Clear |
| Cure | 7 days / RTV |
| Elongation | 150 % |
| Mix Ratio | 100:0.5 |
| Refractive Index | 1.40 |
| Specific Gravity | 1.01 |
| Tack Free Time | 7.5 hours |
| Tensile | 80 psi (0.55 MPa) |
| Work Time | 6.5 hours |
| Comment | Diluent for Tin Condensation CV Silicones |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.