Description
- Two-part, flowable, red RTV silicone
- 100: 0.5 Mix Ratio (Base: Curing Agent)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- As a sealing, caulking, adhesive or potting material in electronic and space applications where high vacuum environments are encountered
- For bonding solar cells to solar array panels
- For applications requiring a broader operating temperature range
Meets or exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤1% and CVCM of ≤0.1%
Product Details
| Property | Average Result |
|---|---|
| Cure System | Alkoxy |
| Viscosity | 55,000 cP (55,000 mPa*s) |
| Lap Shear | 500 psi (3.45 MPa) |
| Durometer | 55 Type A |
| Appearance | Rust Red |
| Cte | |
| Cure | 7 days / RTV |
| Dielectric Strength | 825 volts/mil (32.18 kV/mm) |
| Elongation | 150 % |
| Mix Ratio | 100:0.5 |
| Specific Gravity | 1.49 |
| Tack Free Time | 8 hours |
| Tear | 40 ppi (7.05 kN/m) |
| Tensile | 950 psi (6.55 MPa) |
| Thermal Conductivity | 0.27 W/(mK) |
| Work Time | 3 hours |
| Comment | Pourable RTV Cure, * 1 )Primed Lap Shear 500 psi (3.4 MPa) |
What Is Space Grade?
Discover what it takes to make silicones worthy of the distinction "Space Grade" and the rigorous testing that these materials undergo.