Description
- Two-part, flowable, white RTV silicone
- 100:0.5 Mix Ratio (Base:Curing Agent)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- Use a sealing, caulking, adhesive or potting material in electronic and space applications where high vacuum environments are encountered
- For applications requiring a broader operating temperature range
Meets or exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤1% and CVCM of ≤0.1%
Product Details
| Property | Average Result |
|---|---|
| Cure System | Alkoxy |
| Viscosity | 45,000 cP (45,000 mPa*s) |
| Durometer | 45 Type A |
| Appearance | White |
| Cure | 7 days / RTV |
| Elongation | 200 % |
| Mix Ratio | 100:0.5 |
| Specific Gravity | 1.19 |
| Tack Free Time | 9 hours |
| Tensile | 600 psi (4.14 MPa) |
| Work Time | 4 hours |
| Comment | Low / High Temperature, Flowable |
What Is Space Grade?
Discover what it takes to make silicones worthy of the distinction "Space Grade" and the rigorous testing that these materials undergo.