Description
- Two-part, thixotropic, translucent RTV silicone
- 10:1 Mix Ratio (Part A: Part B)
Applications
- For electronic and space applications requiring low outgassing and minimal volatile condensables to avoid condensation in sensitive devices
- As an embedding or potting compound for environmental protection of electronic assemblies and components
- Especially useful where moderate thixotropy is required
Product Details
| Property | Average Result |
|---|---|
| Cure System | Platinum |
| Durometer | 40 Type A |
| Appearance | Clear |
| Cure | 7 days / RTV |
| Elongation | 220 % |
| Flow Rate | 0.3 in (7.62 mm) |
| Mix Ratio | 10:1 |
| Specific Gravity | 1.06 |
| Tensile | 800 psi (5.52 MPa) |
| Comment | Low Flow |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.