Description
- Two-part, thixotropic, translucent RTV silicone
- 10:1 Mix Ratio (Part A: Part B)
Applications
- For electronic and space applications requiring low outgassing and minimal volatile condensables to avoid condensation in sensitive devices
- As an embedding or potting compound for environmental protection of electronic assemblies and components
- Especially useful where moderate thixotropy is required
Meets or exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤1% and CVCM of ≤0.1%
Product Details
| Property | Average Result |
|---|---|
| Cure System | Platinum |
| Durometer | 40 Type A |
| Appearance | Clear |
| Cure | 7 days / RTV |
| Elongation | 220 % |
| Flow Rate | 0.3 in (7.62 mm) |
| Mix Ratio | 10:1 |
| Specific Gravity | 1.06 |
| Tensile | 800 psi (5.52 MPa) |
| Comment | Low Flow |
High-precision laser performance with ultra-low outgassing silicone
Learn how NuSil collaborated with a leading manufacturer of lasers to improve longevity and stability of space-based lasers.