Description
- Two-part, white silicone system
- Offers a high tear strength, good physical properties and a broad operating temperature range
- Convenient 1:1 mix ratio (Part A: Part B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength
- For use as an adhesive
- For applications requiring a broader operating temperature range
Meets or exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤1% and CVCM of ≤0.1%
Product Details
| Property | Average Result |
|---|---|
| Cure System | Platinum |
| Rheology | Flowable / Self Leveling |
| Work Time | 30 minutes |
| Durometer | 34 Type A |
| Appearance | White |
| Cure | 15 minutes / 150 °C |
| Cure Type | Heat Cure / RTV |
| Dielectric Strength | 955 volts/mil (37.25 kV/mm) |
| Elongation | 343 % |
| Lap Shear | 375 psi (2.6 MPa) |
| Mix Ratio | 1:1 |
| Rheology | Flowable / Self Leveling |
| Specific Gravity | 1.12 |
| Tack Free Time | 5 hours |
| Tensile | 720 psi (5.0 MPa) |
| Viscosity | 50,000 cP (50,000 mPa*s) |
What Is Space Grade?
Discover what it takes to make silicones worthy of the distinction "Space Grade" and the rigorous testing that these materials undergo.