Description
- One-part, black, platinum catalyzed silicone glop-top
- Thixotropic consistency
- Low modulus
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ensures microchip reliability by providing vital protection
- For applications that require precise delivery configurations
Meets or exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤1% and CVCM of ≤0.1%
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 300,000 cP (300,000 mPa*s) |
Extrusion Rate | 0.9 g/minute |
Durometer | 20 Type A |
Appearance | Black |
Cure | 15 minutes / 200 °C |
Cure Rate Scorch | 2.5 minutes |
Elongation | 700 % |
Mix Ratio | 1 PART |
Specific Gravity | 1.15 |
Tear | 50 ppi (8.82 kN/m) |
Tensile | 750 psi (5.17 MPa) |
Volume Resistivity | 1,000,000,000,000,000 ohmcm |
Comment | Thixotropic |
What Is Space Grade?
Discover what it takes to make silicones worthy of the distinction "Space Grade" and the rigorous testing that these materials undergo.