CV-1161
CONTROLLED VOLATILITY PRESSURE SENSITIVE SILICONE ADHESIVE
Description
  • One-part pressure sensitive adhesive
  • Retains adhesive qualities in the presence of minor liquid and particulate contaminants
  • Properties unaffected by normal temperature and pressure variations
Applications
  • For adhering and bonding silicone elastomers, metals, paper, glass, fabric, organic rubbers, some synthetics, and films such as polyester or kapton
  • For applications requiring low outgassing and minimal volatile condensable materials
Meets or exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤1% and CVCM of ≤0.1%
Product Details
Property Average Result
Cure System Non-Curing
Non Volatile Content 50 %
Appearance Translucent
Cure 90 minutes / 150 °C
Post Cure 15 minutes / 150 °C
Viscosity 3,000 cP (3,000 mPa*s)
Comment Clear silicone PSA for adhering and bonding silicone elastomers, metals, paper, glass, fabric, organic rubbers, some synthetics. Excellent for tapes on films such as polyester or kapton.Rlease force of PSA: 7.5 ppi
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.