Description
- One-part, flowable, solvent free silicone
Applications
- For industrial or space applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- As a conformal coating on rigid and flexible circuit boards
- For encapsulating sensitive electronic components such as connectors and switches
- To provide protection against harsh environments while maintaining good dielectric properties
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Viscosity | 7,300 cP (7,300 mPa*s) |
Appearance | Translucent |
Tack Free Time | 50 minutes |
Cure | 7 days / RTV |
Cure System | Oxime |
Mix Ratio | 1 PART |
Refractive Index | 1.43 |
Specific Gravity | 1.01 |
Comment | Soft, very low modulus coating |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.