Description
- One-part silicone dispersion
- Provided as an RTV silicone dispersion in VM&P Naphtha
Applications
- For applications requiring extreme low temperature, low outgassing and minimal volatile condensables under extreme operating conditions
- As a protective overcoating for electronic components and atomic oxygen resistance in space applications
- Provides radiation and thermal stress resistance
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Viscosity | 240 cP (240 mPa*s) |
Non Volatile Content | 60 % |
Appearance | Clear |
Tack Free Time | 50 minutes |
Cure | 7 days / RTV |
Cure System | Oxime |
Mix Ratio | 1 PART |
Refractive Index | 1.43 |
Specific Gravity | 1 |
Comment | 60% Solids, Atomic Oxygen Protective Overcoat |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.