Description
- Volatile carrier for silicone elastomer gels, gum blends, and fluids
- Compatible with a range of cosmetic ingredients
- Further reduces the non-volatile content of silicone elastomer gels, gum blends, and fluids
- Delivers a silky feel within skin care applications
- Delivers enhanced spreadability and emolliency within skin care formulations
Applications
- Moisturization
- Anti-Aging/Anti-Wrinkle
- Sunscreen, After-Sun, and Self-Tanners
- Skin Repair Serums
- Skin Tone Repair
- Men's Grooming Color
- Cosmetics
Product Details
Property | Average Result |
---|---|
INCI Name | Cyclohexasiloxane |
Viscosity | 7 cSt (6.77 mPa*s) |
Appearance | Clear |
Specific Gravity | 0.97 |
D4 Content | <0.05 % |
Refractive Index | 1.40 |
Comment | Pure Volatile Silicone Gas Chromatography: (D4):<0.05% (D5):<0.05% (D6):<99.5% |