Description
- CFF-3401 offers transfer and wash-off resistance in color cosmetics
- Enhanced hydrophobic and oleophobic properties
- Acts as an ingredient to deliver moisturizing properties
- Unique fluoro chemistry designed for skin care and color cosmetic applications
- Enhanced substanstivity to the skin
- Enhanced spreadability in comparison to standard Dimethicone
- Reduces tackiness in formulations
- Low refractive index
- Enhanced pigment wetting in color cosmetics
Applications
- Moisturization
- Anti-Aging/Anti-Wrinkle
- Sunscreen, After- Sun, and Self- Tanners
- Skin Repair Serums
- Skin Tone Repair
- Men’s Grooming
- Color Cosmetics
Product Details
Property | Average Result |
---|---|
INCI Name | Trifluoropropyl Dimethicone |
D4 Content | <0.05 % |
Viscosity | 350 cP / 1,000 cP / 25,000 cP (350 mPa*s / 1,000 mPa*s / 25,000 mPa*s) |
Appearance | Clear |
Refractive Index | 1.38 |
Specific Gravity | 1.21 |
Comment | 60 Mol% Fluorosilicone |